Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ITGA6 Partial ORF (NP_000201, 24 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003655Q01L

 View more versions of this product

Catalog No. 89-949-268


Add to cart

Description

Description

The ITGA6 protein product is the integrin alpha chain alpha 6. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: FNLDTREDNVIRKYGDPGSLFGFSLAMHWQLQPEDKRLLLVGAPRAEALPLQRANRTGGLYSCDITARGPCTRIEFDNDADPTSESKEDQWMGVTVQSQGPGGKVVTCAH
Specifications

Specifications

NP_000201
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FNLDTREDNVIRKYGDPGSLFGFSLAMHWQLQPEDKRLLLVGAPRAEALPLQRANRTGGLYSCDITARGPCTRIEFDNDADPTSESKEDQWMGVTVQSQGPGGKVVTCAH
RUO
ITGA6
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3655
ITGA6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD49f|DKFZp686J01244|FLJ18737|ITGA6B|VLA-6
ITGA6
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit