Learn More
Abnova™ Human ITK (NM_005546, 1 a.a.- 620 a.a.) Full-length Recombinant Protein expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4707
Catalog No. 89-949-347
Add to cart
Description
Description
This gene encodes an intracellular tyrosine kinase expressed in T-cells. The protein contains both SH2 and SH3 domains which are often found in intracellular kinases. It is thought to play a role in T-cell proliferation and differentiation. [provided by RefSeq]
Specifications
Specifications
NM_005546
0.073 ug/uL
Liquid
3702
ITK (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
ITK
11pmol/μg x min
Yes
Insect cell (Sf9) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
101.727kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
EMT|LYK|MGC126257|MGC126258|PSCTK2
ITK
Insect
None