Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ITSN2 Partial ORF (NP_006268, 471 a.a. - 571 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00050618Q01L

 View more versions of this product

Catalog No. 89-949-378


Add to cart

Description

Description

This gene encodes a cytoplasmic protein which contains SH3 domains. This protein is a member of a family of proteins involved in clathrin-mediated endocytosis. Intersectin 2 is thought to regulate the formation of clathrin-coated vesicles and also may function in the induction of T cell antigen receptor (TCR) endocytosis. Alternatively spliced transcript variants have been found for this gene that encode three distinct isoforms. Additional variants have been found but their full length nature has not been determined. [provided by RefSeq]

Sequence: NQKNREQEEIVRLNSKKKNLHLELEALNGKHQQISGRLQDVRLKKQTQKTELEVLDKQCDLEIMEIKQLQQELQEYQNKLIYLVPEKQLLNERIKNMQFSN
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NQKNREQEEIVRLNSKKKNLHLELEALNGKHQQISGRLQDVRLKKQTQKTELEVLDKQCDLEIMEIKQLQQELQEYQNKLIYLVPEKQLLNERIKNMQFSN
RUO
ITSN2
Wheat Germ (in vitro)
GST
NP_006268
Liquid
50618
ITSN2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1256/PRO2015/SH3D1B/SH3P18/SWA/SWAP
ITSN2
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit