Learn More
Abnova™ Human JAK2 (NM_004972.2, 717 a.a. - 1132 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4708
Catalog No. 89-949-398
Add to cart
Description
Description
This gene product is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways. It has been found to be constituitively associated with the prolactin receptor and is required for responses to gamma interferon. Mice that do not express an active protein for this gene exhibit embryonic lethality associated with the absence of definitive erythropoiesis. [provided by RefSeq]
Specifications
Specifications
NM_004972.2
0.091 ug/uL
Liquid
3717
JAK2 (Human) Recombinant Protein
GST affinity chromatography
100 ug
RUO
JAK2
24pmol/μg x min
Yes
Insect cell (Sf9) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
78.512kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
JTK10
JAK2
Insect
GST-His