Learn More
Abnova™ Human JAK3 (NP_000206.1, 781 a.a. - 1124 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4709
Catalog No. 89-949-405
Add to cart
Description
Description
The protein encoded by this gene is a member of the Janus kinase (JAK) family of tyrosine kinases involved in cytokine receptor-mediated intracellular signal transduction. It is predominantly expressed in immune cells and transduces a signal in response to its activation via tyrosine phosphorylation by interleukin receptors. Mutations in this gene are associated with autosomal SCID (severe combined immunodeficiency disease). [provided by RefSeq]
Specifications
Specifications
NP_000206.1
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
68.379kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
JAK-3|JAK3_HUMAN|JAKL|L-JAK|LJAK
JAK3
Insect
GST-His
Insect cell (Sf9) expression system
Liquid
3718
JAK3 (Human) Recombinant Protein
GST-Affinity Chromatography
100 ug
RUO
JAK3
5pmol/μg x min
Yes