Learn More
Raybiotech IncSupplier Diversity Partner Human Junctional Adhesion Molecule B (JAM2 / VE-JAM / CD322) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHJAMB1
For Serum, Plasma, and Cell Culture Supernatants. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Junctional Adhesion Molecule B (JAM2 / VE-JAM / CD322)
Catalog No. 50-194-9474