Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KAAG1 (NP_851854, 1 a.a. - 84 a.a.) Full-length Recombinant Protein with His tag

Used for SDS-PAGE

Manufacturer:  Abnova™ P3429

Catalog No. 89-949-472


Add to cart

Description

Description

Sequence: MGSSHHHHHHSSGLVPRGSHMDDDAAPRVEGVPVAVHKHALHDGLRQVAGPGAAAAHLPRWPPPQLAASRREAPPLSQRPHRTQGAGSPPETNEKLTNPQVKEK
Specifications

Specifications

Escherichia coli expression system
0.5 mg/mL
Liquid
353219
KAAG1 (Human) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 15% SDS-PAGE
MGSSHHHHHHSSGLVPRGSHMDDDAAPRVEGVPVAVHKHALHDGLRQVAGPGAAAAHLPRWPPPQLAASRREAPPLSQRPHRTQGAGSPPETNEKLTNPQVKEK
RUO
KAAG1
Escherichia coli
His
NP_851854
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
11.1kDa
Escherichia coli expression system
>90% by SDS-PAGE
10 ug
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
MGC78738|RU2AS
KAAG1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit