Learn More
Novus Biologicals Human Kallikrein 6/Neurosin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP262156
Description
The Human Kallikrein 6/Neurosin ELISA Kit (Colorimetric) measures human Kallikrein 6/Neurosin in serum.Specifications
ELISA Kit (Colorimetric) | |
No cross-reactivity with other serine proteases was detected for this assay | |
5653 | |
Kallikrein 6/Neurosin | |
1 Kit | |
1 ng/ml |
5-280 ng/ml | |
ELISA | |
Store at 4°C | |
Human | |
Serum | |
KLK6 |