Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Kallikrein 7 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP262157
Description
The Human Kallikrein 7 ELISA Kit (Colorimetric) measures human Kallikrein 7 in serum.Specifications
35-560 ng/ml | |
No cross-reactivity with other serine proteases was detected for this assay. | |
5650 | |
Human | |
1 Kit | |
Store at 4°C |
0.5 ng/ml | |
ELISA | |
KLK7 | |
ELISA Kit (Colorimetric) | |
Serum | |
Kallikrein 7 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title