Learn More
Novus Biologicals Human Kallikrein 8/Neuropsin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP262176
Description
The Human Kallikrein 8/Neuropsin ELISA Kit (Colorimetric) measures human Kallikrein 8/Neuropsin in serum.Specifications
ELISA Kit (Colorimetric) | |
No cross-reactivity with other serine proteases was detected for this assay. | |
11202 | |
Kallikrein 8/Neuropsin | |
1 Kit | |
0.1 ng/ml |
35-560 ng/ml | |
ELISA | |
Store at 4°C | |
Human | |
Serum | |
KLK8 |