Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KCNAB3 Full-length ORF (NP_004723.2, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009196P01S

 View more versions of this product

Catalog No. 89-949-505


Add to cart

Description

Description

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. This member and the KCNA5 gene product assemble into a heteromultimeric A-type channel that inactivates completely and is significantly faster than other A-type Kv channels. [provided by RefSeq]

Sequence: MQVSIACTEQNLRSRSSEDRLCGPRPGPGGGNGGPAGGGHGNPPGGGGSGPKARAALVPRPPAPAGALRESTGRGTGMKYRNLGKSGLRVSCLGLGTWVTFGSQISDETAEDVLTVAYEHGVNLFDTAEVYAAGKAERTLGNILKSKGWRRSSYVITTKIFWGGQAETERGLSRKHIIEGLRGSLERLQLGYVDIVFANRSDPNCPMEEIVRAMTYVINQGLALYWGTSRWGAAEIMEAYSMARQFNLIPPVCEQAEHHLFQREKVEMQLPELYHKIGVGSVTWYPLACGLITSKYDGRVPDTCRASIKGYQWLKDKVQSEDGKKQQAKVMDLLPVAHQLGCTVAQLAIAWCLRSEGVSSVLLGVSSAEQLIEHLGALQVLSQLTPQTVMEIDGLLGNKPHSKK
Specifications

Specifications

NP_004723.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKR6A9|KCNA3.1B|KCNA3B|KV-BETA-3|MGC116886
KCNAB3
Yes
wheat germ expression system
Liquid
9196
KCNAB3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MQVSIACTEQNLRSRSSEDRLCGPRPGPGGGNGGPAGGGHGNPPGGGGSGPKARAALVPRPPAPAGALRESTGRGTGMKYRNLGKSGLRVSCLGLGTWVTFGSQISDETAEDVLTVAYEHGVNLFDTAEVYAAGKAERTLGNILKSKGWRRSSYVITTKIFWGGQAETERGLSRKHIIEGLRGSLERLQLGYVDIVFANRSDPNCPMEEIVRAMTYVINQGLALYWGTSRWGAAEIMEAYSMARQFNLIPPVCEQAEHHLFQREKVEMQLPELYHKIGVGSVTWYPLACGLITSKYDGRVPDTCRASIKGYQWLKDKVQSEDGKKQQAKVMDLLPVAHQLGCTVAQLAIAWCLRSEGVSSVLLGVSSAEQLIEHLGALQVLSQLTPQTVMEIDGLLGNKPHSKK
RUO
KCNAB3
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit