Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KCNH7 Partial ORF (NP_775185.1, 132 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00090134Q01L

 View more versions of this product

Catalog No. 89-949-526


Add to cart

Description

Description

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. There are at least two alternatively spliced transcript variants derived from this gene and encoding distinct isoforms. [provided by RefSeq]

Sequence: VTDNENAATPERVNPILPIKTVNRKFFGFKFPGLRVLTYRKQSLPQEDPDVVVIDSSKHSDDSVAMKHFKSPTKESCSPSEADDTKALIQPSKCSPLVN
Specifications

Specifications

NP_775185.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VTDNENAATPERVNPILPIKTVNRKFFGFKFPGLRVLTYRKQSLPQEDPDVVVIDSSKHSDDSVAMKHFKSPTKESCSPSEADDTKALIQPSKCSPLVN
RUO
KCNH7
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
90134
KCNH7 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ERG3|HERG3|Kv11.3|MGC45986
KCNH7
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit