Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KCNN3 Full-length ORF (NP_740752.1, 1 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003782P02L

 View more versions of this product

Catalog No. 89-949-567


Add to cart

Description

Description

Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. The protein encoded by this gene is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits. This gene contains two CAG repeat regions in the coding sequence. It was thought that expansion of one or both of these repeats could lead to an increased susceptibility to schizophrenia or bipolar disorder, but studies indicate that this is probably not the case. This gene is a member of the KCNN family of potassium channel genes. Two transcript variants encoding two different isoforms have been found for this gene. One of the variants lacks the CAG repeat regions. [provided by RefSeq]

Sequence: MERPIKDSMFSLALKCLISLSTIILLGLIIAYHTREVQLFVIDNGADDWRIAMTYERILYISLEMLVCAIHPIPGEYKFFWTARLAFSYTPSRAEADVDIILSIPMFLRLYLIARVMLLHSKLFTDASSRSIGALNKINFNTRFVMKTLMTICPGTVLLVFSISLWIIAAWTVRVCERYHDQQDVTSNFLGAMWLISITFLSIGYGDMVPHTYCGKGVCLLTGIMGAGCTALVVAVVARKLELTKAEKHVHNFMMDTQLTKRIKNAAANVLRETWLIYKHTKLLKKIDHAKVRKHQRKFLQAIHQLRSVKMEQRKLSDQANTLVDLSKMQNVMYDLITELNDRSEDLEKQIGSLESKLEHLTASFNSLPLLIADTLRQQQQQLLSAIIEARGVSVAVGTTHTPISDSPIGVSSTSFPTPYTSSSSC
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KCa2.3|SK3|SKCA3|hSK3
KCNN3
Yes
NP_740752.1
Liquid
3782
KCNN3 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERPIKDSMFSLALKCLISLSTIILLGLIIAYHTREVQLFVIDNGADDWRIAMTYERILYISLEMLVCAIHPIPGEYKFFWTARLAFSYTPSRAEADVDIILSIPMFLRLYLIARVMLLHSKLFTDASSRSIGALNKINFNTRFVMKTLMTICPGTVLLVFSISLWIIAAWTVRVCERYHDQQDVTSNFLGAMWLISITFLSIGYGDMVPHTYCGKGVCLLTGIMGAGCTALVVAVVARKLELTKAEKHVHNFMMDTQLTKRIKNAAANVLRETWLIYKHTKLLKKIDHAKVRKHQRKFLQAIHQLRSVKMEQRKLSDQANTLVDLSKMQNVMYDLITELNDRSEDLEKQIGSLESKLEHLTASFNSLPLLIADTLRQQQQQLLSAIIEARGVSVAVGTTHTPISDSPIGVSSTSFPTPYTSSSSC
RUO
KCNN3
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit