Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KCNV1 Full-length ORF (AAH28739.1, 1 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00027012P01S

 View more versions of this product

Catalog No. 89-949-583


Add to cart

Description

Description

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium voltage-gated channel subfamily V. This protein is essentially present in the brain, and its role might be to inhibit the function of a particular class of outward rectifier potassium channel types. [provided by RefSeq]

Sequence: MPSSGRALLDSPLDSGSLTSLGSSVFCSEGEGEPLALGDCFTVNVGGSRFVLSQQALSCFPHTRLGKLAVVVASYRRPGALAAVPSPLELCDDANPVDNEYFFDRSSQAFRYVLHYYRTGRLHVMEQLCALSFLQEIQYWGIDELSIDSCCRDRYFRRKELSETLDFKKDTEDQESQHESEQDFSQGPCPTVRQKLWNILEKPGSSTAARIFGVISIIFVVVSIINMALMSAELSWLDLQLLEILEYVCISWFTGEFVLRFLCVRDRCRFLRKVPNIIDLLAILPFYITLLVESLSGSQTTQELENVGRIVQVLRLLRALRMLKLGRHSTGLRSLGMTITQCYEEVGLLLLFLSVGISIFSTVEYFAEQSIPDTTFTSVPCAWWWATTSMTTVGYGDIRPDTTTGKIVAFMCILSGILVLALPIAIINDRFSACYFTLKLKEAAVRQREALKKLTKNIATDSYISVNLRDVYARSIMEMLRLKGRERASTRSSGGDDFWF
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
82.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HNKA|KCNB3|KV2.3|KV8.1
KCNV1
Yes
AAH28739.1
Liquid
27012
KCNV1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPSSGRALLDSPLDSGSLTSLGSSVFCSEGEGEPLALGDCFTVNVGGSRFVLSQQALSCFPHTRLGKLAVVVASYRRPGALAAVPSPLELCDDANPVDNEYFFDRSSQAFRYVLHYYRTGRLHVMEQLCALSFLQEIQYWGIDELSIDSCCRDRYFRRKELSETLDFKKDTEDQESQHESEQDFSQGPCPTVRQKLWNILEKPGSSTAARIFGVISIIFVVVSIINMALMSAELSWLDLQLLEILEYVCISWFTGEFVLRFLCVRDRCRFLRKVPNIIDLLAILPFYITLLVESLSGSQTTQELENVGRIVQVLRLLRALRMLKLGRHSTGLRSLGMTITQCYEEVGLLLLFLSVGISIFSTVEYFAEQSIPDTTFTSVPCAWWWATTSMTTVGYGDIRPDTTTGKIVAFMCILSGILVLALPIAIINDRFSACYFTLKLKEAAVRQREALKKLTKNIATDSYISVNLRDVYARSIMEMLRLKGRERASTRSSGGDDFWF
RUO
KCNV1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit