Learn More
Abnova™ Human KDELC1 Full-length ORF (NP_076994.1, 1 a.a. - 502 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00079070P01S
Description
This gene encodes a protein product localized to the lumen of the endoplasmic reticulum. As a member of the endoplasmic reticulum protein family the encoded protein contains a Lys-Asp-Glu-Leu or KDEL motif located at the extreme C-terminus which prevents all endoplasmic reticulum resident proteins from being secreted. Proteins carrying this motif are bound by a receptor in the Golgi apparatus so that the receptor-ligand complex returns to the endoplasmic reticulum. A processed non-transcribed pseudogene located in an intron of a sodium transporter gene on chromosome 5 has been defined for this gene. [provided by RefSeq]
Sequence: MFGTLLLYCFFLATVPALAETGGERQLSPEKSEIWGPGLKADVVLPARYFYIQAVDTSGNKFTSSPGEKVFQVKVSAPEEQFTRVGVQVLDRKDGSFIVRYRMYASYKNLKVEVKFQGQHVAKSPYILKGPVYHENCDCPLQDSAAWLREMNCPETIAQIQRDLAHFPAVDPEKIAVEIPKRFGQRQSLCHYTLKDNKVYIKTHGEHVGFRIFMDAILLSLTRKVKMPDVELFVNLGDWPLEKKKSNSNIHPIFSWCGSTDSKDIVMPTYDLTDSVLETMGRVSLDMMSVQANTGPPWESKNSTAVWRGRDSRKERLELVKLSRKHPELIDAAFTNFFFFKHDENLYGPIVKHISFFDFFKHKYQINIDGTVAAYRLPYLLVGDSVVLKQDSIYYEHFYNELQPWKHYIPVKSNLSDLLEKLKWAKDHDEEAKKIAKAGQEFARNNLMGDDIFCYYFKLFQEYANLQVSEPQIREGMKRVEPQTEDDLFPCTCHRKKTKDELSpecifications
NP_076994.1 | |
Liquid | |
79070 | |
KDELC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFGTLLLYCFFLATVPALAETGGERQLSPEKSEIWGPGLKADVVLPARYFYIQAVDTSGNKFTSSPGEKVFQVKVSAPEEQFTRVGVQVLDRKDGSFIVRYRMYASYKNLKVEVKFQGQHVAKSPYILKGPVYHENCDCPLQDSAAWLREMNCPETIAQIQRDLAHFPAVDPEKIAVEIPKRFGQRQSLCHYTLKDNKVYIKTHGEHVGFRIFMDAILLSLTRKVKMPDVELFVNLGDWPLEKKKSNSNIHPIFSWCGSTDSKDIVMPTYDLTDSVLETMGRVSLDMMSVQANTGPPWESKNSTAVWRGRDSRKERLELVKLSRKHPELIDAAFTNFFFFKHDENLYGPIVKHISFFDFFKHKYQINIDGTVAAYRLPYLLVGDSVVLKQDSIYYEHFYNELQPWKHYIPVKSNLSDLLEKLKWAKDHDEEAKKIAKAGQEFARNNLMGDDIFCYYFKLFQEYANLQVSEPQIREGMKRVEPQTEDDLFPCTCHRKKTKDEL | |
RUO | |
KDELC1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EP58/KDEL1/MGC5302 | |
KDELC1 | |
Yes | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.