Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KEL Full-length ORF (NP_000411.1, 1 a.a. - 732 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003792P01S

 View more versions of this product

Catalog No. 89-949-622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases. [provided by RefSeq]

Sequence: MEGGDQSEEEPRERSQAGGMGTLWSQESTPEERLPVEGSRPWAVARRVLTAILILGLLLCFSVLLFYNFQNCGPRPCETSVCLDLRDHYLASGNTSVAPCTDFFSFACGRAKETNNSFQELATKNKNRLRRILEVQNSWHPGSGEEKAFQFYNSCMDTLAIEAAGTGPLRQVIEELGGWRISGKWTSLNFNRTLRLLMSQYGHFPFFRAYLGPHPASPHTPVIQIDQPEFDVPLKQDQEQKIYAQIFREYLTYLNQLGTLLGGDPSKVQEHSSLSISITSRLFQFLRPLEQRRAQGKLFQMVTIDQLKEMAPAIDWLSCLQATFTPMSLSPSQSLVVHDVEYLKNMSQLVEEMLLKQRDFLQSHMILGLVVTLSPALDSQFQEARRKLSQKLRELTEQPPMPARPRWMKCVEETGTFFEPTLAALFVREAFGPSTRSAAMKLFTAIRDALITRLRNLPWMNEETQNMAQDKVAQLQVEMGASEWALKPELARQEYNDIQLGSSFLQSVLSCVRSLRARIVQSFLQPHPQHRWKVSPWDVNAYYSVSDHVVVFPAGLLQPPFFHPGYPRAVNFGAAGSIMAHELLHIFYQLLLPGGCLACDNHALQEAHLCLKRHYAAFPLPSRTSFNDSLTFLENAADVGGLAIALQAYSKRLLRHHGETVLPSLDLSPQQIFFRSYAQVMCRKPSPQDSHDTHSPPHLRVHGPLSSTPAFARYFRCARGALLNPSSRCQLW
Specifications

Specifications

NP_000411.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
109.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD238|ECE3
KEL
Yes
wheat germ expression system
Liquid
3792
KEL (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEGGDQSEEEPRERSQAGGMGTLWSQESTPEERLPVEGSRPWAVARRVLTAILILGLLLCFSVLLFYNFQNCGPRPCETSVCLDLRDHYLASGNTSVAPCTDFFSFACGRAKETNNSFQELATKNKNRLRRILEVQNSWHPGSGEEKAFQFYNSCMDTLAIEAAGTGPLRQVIEELGGWRISGKWTSLNFNRTLRLLMSQYGHFPFFRAYLGPHPASPHTPVIQIDQPEFDVPLKQDQEQKIYAQIFREYLTYLNQLGTLLGGDPSKVQEHSSLSISITSRLFQFLRPLEQRRAQGKLFQMVTIDQLKEMAPAIDWLSCLQATFTPMSLSPSQSLVVHDVEYLKNMSQLVEEMLLKQRDFLQSHMILGLVVTLSPALDSQFQEARRKLSQKLRELTEQPPMPARPRWMKCVEETGTFFEPTLAALFVREAFGPSTRSAAMKLFTAIRDALITRLRNLPWMNEETQNMAQDKVAQLQVEMGASEWALKPELARQEYNDIQLGSSFLQSVLSCVRSLRARIVQSFLQPHPQHRWKVSPWDVNAYYSVSDHVVVFPAGLLQPPFFHPGYPRAVNFGAAGSIMAHELLHIFYQLLLPGGCLACDNHALQEAHLCLKRHYAAFPLPSRTSFNDSLTFLENAADVGGLAIALQAYSKRLLRHHGETVLPSLDLSPQQIFFRSYAQVMCRKPSPQDSHDTHSPPHLRVHGPLSSTPAFARYFRCARGALLNPSSRCQLW
RUO
KEL
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit