Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KGF/FGF-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | KGF/FGF-7 |
---|---|
Clone | 29568 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
KGF/FGF-7 Monoclonal specifically detects KGF/FGF-7 in Human samples. It is validated for Western Blot.Specifications
KGF/FGF-7 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) | |
FGF7 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
29568 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P21781 | |
2252 | |
E. coli-derived recombinant human KGF Cys32-Thr194 Accession # P21781 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human KGF in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) FGF-3, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF-9, rhFGF-10, rhFGF-11, rhFGF-13, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF-19, rhFGF acidic, rhFGF basic, rmFGF-8b, rmFGF-8c, or rmFGF-15. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title