Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIAA1586 Full-length ORF (NP_065982.1, 1 a.a. - 787 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057691P01

Catalog No. 89-949-812


Add to cart

Description

Description

Sequence: MGDPGSEIIESVPPAGPEASESTTDENEDDIQFVSEGPSRPVLEYIDLVCGDDENPSAYYSDILFPKMPKRQGDFLHFLNVKKVKTDTENNEVSKNHCRLSKAKEPHFEYIEQPIIEEKPSLSSKKEIDNLVLPDCWNEKQAFMFTEQYKWLEIKEGKLGCKDCSAVRHLGSKAEKHVHVSKEWIAYLVTPNGSNKTTRQASLRKKIREHDVSKAHGKIQDLLKESTNDSICNLVHKQNNKNIDATVKVFNTVYSLVKHNRPLSDIEGARELQEKNGEVNCLNTRYSATRIAEHIAKEMKMKIFKNIIEENAKICIIIDEASTVSKKTTLVIYLQCTIQSAPAPVMLFVALKELVSTIAECIVNTLLTTLNDCGFTNEYLKANLIAFCSDGANTILGRKSGVATKLLENFPEIIIWNCLNHRLQLSLDDSISEIKQINHLKIFIDKIYSIYHQPNKNQTKLLGTVAKELETEIIKIGRVMGPRWAACSLQAATAVWHAYPILYMHFSHSYSGLAKRLANINFLQDLALMIDILEEFSVLSTALQSRSTNIKKAQKLIKRTIRALENLKIGTGKYESQIEDLIKSDKFKDIPFNKNNKFNALPRSILLDNIIQHMNLRLLSDRNHEDIFNYFDLLEPSTWPYEEITSPWIAGEKTLFHLCKILKYEVDLNDFREFVNNNIKSNNVSIPTTIYKAKKIVSTIAINSAEAERGFNLMNIICTRVRNSLTIDHVSDLMTINLLGKELADWDATPFVKSWSNCNHRLATDTRVRQKSTKVFHENQLAIWNLK
Specifications

Specifications

NP_065982.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
116.1kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ30162
KIAA1586
Yes
wheat germ expression system
Liquid
57691
KIAA1586 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGDPGSEIIESVPPAGPEASESTTDENEDDIQFVSEGPSRPVLEYIDLVCGDDENPSAYYSDILFPKMPKRQGDFLHFLNVKKVKTDTENNEVSKNHCRLSKAKEPHFEYIEQPIIEEKPSLSSKKEIDNLVLPDCWNEKQAFMFTEQYKWLEIKEGKLGCKDCSAVRHLGSKAEKHVHVSKEWIAYLVTPNGSNKTTRQASLRKKIREHDVSKAHGKIQDLLKESTNDSICNLVHKQNNKNIDATVKVFNTVYSLVKHNRPLSDIEGARELQEKNGEVNCLNTRYSATRIAEHIAKEMKMKIFKNIIEENAKICIIIDEASTVSKKTTLVIYLQCTIQSAPAPVMLFVALKELVSTIAECIVNTLLTTLNDCGFTNEYLKANLIAFCSDGANTILGRKSGVATKLLENFPEIIIWNCLNHRLQLSLDDSISEIKQINHLKIFIDKIYSIYHQPNKNQTKLLGTVAKELETEIIKIGRVMGPRWAACSLQAATAVWHAYPILYMHFSHSYSGLAKRLANINFLQDLALMIDILEEFSVLSTALQSRSTNIKKAQKLIKRTIRALENLKIGTGKYESQIEDLIKSDKFKDIPFNKNNKFNALPRSILLDNIIQHMNLRLLSDRNHEDIFNYFDLLEPSTWPYEEITSPWIAGEKTLFHLCKILKYEVDLNDFREFVNNNIKSNNVSIPTTIYKAKKIVSTIAINSAEAERGFNLMNIICTRVRNSLTIDHVSDLMTINLLGKELADWDATPFVKSWSNCNHRLATDTRVRQKSTKVFHENQLAIWNLK
RUO
KIAA1586
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit