Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIF12 Full-length ORF (NP_612433.1, 1 a.a. - 513 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00113220P01

Catalog No. 89-949-864


Add to cart

Description

Description

KIF12 is a member of the kinesin superfamily of microtubule-associated molecular motors (see MIM 148760) that play important roles in intracellular transport and cell division (Nakagawa et al., 1997 [PubMed 9275178]).[supplied by OMIM]

Sequence: MQRTFAWLLDRVQHLGAPVTLRASYLEIYNEQVRDLLSLGSPRPLPVRWNKTRGFYVEQLRVVEFGSLEALMELLQTGLSRRRNSAHTLNQASSRSHALLTLYISRQTAQQMPSVDPGEPPVGGKLCFVDLAGSEKVAATGSRGELMLEANSINRSLLALGHCISLLLDPQRKQSHIPFRDSKLTKLLADSLGGRGVTLMVACVSPSAQCLPETLSTLRYASRAQRVTTRPQAPKSPVAKQPQRLETEMLQLQEENRRLQFQLDQMDCKASGLSGARVAWAQRNLYGMLQEFMLENERLRKEKSQLQNSRDLAQNEQRILAQQVHALERRLLSACYHHQQGPGLTPPCPCLMAPAPPCHALPPLYSCPCCHICPLCRVPLAHWACLPGEHHLPQVLDPEASGGRPPSARPPPWAPPCSPGSAKCPRERSHSDWTQTRVLAEMLTEEEVVPSAPPLPVRPPKTSPGLRGGAGVPNLAQRLEALRDQIGSSLRRGRSQPPCSEGARSPGQVLPPH
Specifications

Specifications

NP_612433.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
83kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RP11-56P10.3
KIF12
Yes
wheat germ expression system
Liquid
113220
KIF12 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQRTFAWLLDRVQHLGAPVTLRASYLEIYNEQVRDLLSLGSPRPLPVRWNKTRGFYVEQLRVVEFGSLEALMELLQTGLSRRRNSAHTLNQASSRSHALLTLYISRQTAQQMPSVDPGEPPVGGKLCFVDLAGSEKVAATGSRGELMLEANSINRSLLALGHCISLLLDPQRKQSHIPFRDSKLTKLLADSLGGRGVTLMVACVSPSAQCLPETLSTLRYASRAQRVTTRPQAPKSPVAKQPQRLETEMLQLQEENRRLQFQLDQMDCKASGLSGARVAWAQRNLYGMLQEFMLENERLRKEKSQLQNSRDLAQNEQRILAQQVHALERRLLSACYHHQQGPGLTPPCPCLMAPAPPCHALPPLYSCPCCHICPLCRVPLAHWACLPGEHHLPQVLDPEASGGRPPSARPPPWAPPCSPGSAKCPRERSHSDWTQTRVLAEMLTEEEVVPSAPPLPVRPPKTSPGLRGGAGVPNLAQRLEALRDQIGSSLRRGRSQPPCSEGARSPGQVLPPH
RUO
KIF12
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit