Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIF21B Partial ORF (XP_371332, 1183 a.a. - 1282 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023046Q01S

 View more versions of this product

Catalog No. 89-949-879


Add to cart

Description

Description

KIF21B belongs to a family of plus end-directed kinesin (see MIM 600025) motor proteins. Neurons use kinesin and dynein (see MIM 600112) microtubule-dependent motor proteins to transport essential cellular components along axonal and dendritic microtubules.[supplied by OMIM]

Sequence: VSRTVSLPTRGSTFPRQSRATETSPLTRRKSYDRGQPIRSTDVGFTPPSSPPTRPRNDRNVFSRLTSNQSQGSALDKSDDSDSSLSEVLRGIISPVGGAK
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VSRTVSLPTRGSTFPRQSRATETSPLTRRKSYDRGQPIRSTDVGFTPPSSPPTRPRNDRNVFSRLTSNQSQGSALDKSDDSDSSLSEVLRGIISPVGGAK
RUO
KIF21B
Wheat Germ (in vitro)
GST
XP_371332
Liquid
23046
KIF21B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16314
KIF21B
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit