Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIFAP3 Full-length ORF (ABM85040.1, 1 a.a. - 792 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00022920P01L

 View more versions of this product

Catalog No. 89-949-907


Add to cart

Description

Description

The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. [provided by RefSeq]

Sequence: MQGEDARYLKRKVKGGNIDVHPSEKALIVHYEVEATILGEMGDPMLGERKECQKIIRLKSLNANTDITSLARKVVEECKLIHPSKLNEVEQLLYYLQNRRDSLSGKEKKEKSSKPKDPPPFEGMEIDEVANINDMDEYIELLYEDIPDKVRGSALILQLARNPDNLEELLLNETALGALARVLREDWKQSVELATNIIYIFFCFSSFSQFHGLITHYKIGALCMNIIDHELKRHELWQEELSKKKKAVDEDPENQTLRKDYEKTFKKYQGLVVKQEQLLRVALYLLLNLAEDTRTELKMRNKNIVHMLVKALDRDNFELLILVVSFLKKLSIFMENKNDMVEMDIVEKLVKMIPCEHEDLLNITLRLLLNLSFDTGLRNKMVQVGLLPKLTALLGNDNYKQIAMCVLYHISMDDRFKSMFAYTDCIPQLMKMLFECSDERIDLELISFCINLAANKRNVQLICEGNGLKMLMKRALKFKDPLLMKMIRNISQHDGPTKNLFIDYVGDLAAQISNDEEEEFVIECLGTLANLTIPDLDWELVLKEYKLVPYLKDKLKPGAAEDDLVLEVVIMIGTVSMDDSCAALLAKSGIIPALIELLNAQQEDDEFVCQIIYVFYQMVFHQATRDVIIKETQAPAYLIDLMHDKNNEIRKVCDNTLDIIAEYDEEWAKKIQSEKFRWHNSQWLEMVESRQMDESEQYLYGDDRIEPYIHEGDILERPDLFYNSDGLIASEGAISPDFFNDYHLQNGDVVGQHSFPGSLGMDGFGQPVGILGRPATAYGFRPDEPYYYGYGS
Specifications

Specifications

ABM85040.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
113.52kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22818|KAP3|SMAP|Smg-GDS|dJ190I16.1
KIFAP3
Yes
wheat germ expression system
Liquid
22920
KIFAP3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MQGEDARYLKRKVKGGNIDVHPSEKALIVHYEVEATILGEMGDPMLGERKECQKIIRLKSLNANTDITSLARKVVEECKLIHPSKLNEVEQLLYYLQNRRDSLSGKEKKEKSSKPKDPPPFEGMEIDEVANINDMDEYIELLYEDIPDKVRGSALILQLARNPDNLEELLLNETALGALARVLREDWKQSVELATNIIYIFFCFSSFSQFHGLITHYKIGALCMNIIDHELKRHELWQEELSKKKKAVDEDPENQTLRKDYEKTFKKYQGLVVKQEQLLRVALYLLLNLAEDTRTELKMRNKNIVHMLVKALDRDNFELLILVVSFLKKLSIFMENKNDMVEMDIVEKLVKMIPCEHEDLLNITLRLLLNLSFDTGLRNKMVQVGLLPKLTALLGNDNYKQIAMCVLYHISMDDRFKSMFAYTDCIPQLMKMLFECSDERIDLELISFCINLAANKRNVQLICEGNGLKMLMKRALKFKDPLLMKMIRNISQHDGPTKNLFIDYVGDLAAQISNDEEEEFVIECLGTLANLTIPDLDWELVLKEYKLVPYLKDKLKPGAAEDDLVLEVVIMIGTVSMDDSCAALLAKSGIIPALIELLNAQQEDDEFVCQIIYVFYQMVFHQATRDVIIKETQAPAYLIDLMHDKNNEIRKVCDNTLDIIAEYDEEWAKKIQSEKFRWHNSQWLEMVESRQMDESEQYLYGDDRIEPYIHEGDILERPDLFYNSDGLIASEGAISPDFFNDYHLQNGDVVGQHSFPGSLGMDGFGQPVGILGRPATAYGFRPDEPYYYGYGS
RUO
KIFAP3
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit