Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIFAP3 Full-length ORF (ABM85040.1, 1 a.a. - 792 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name KIFAP3 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 113.52kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-949-907
View Documents
Abnova™
H00022920P01L
25 ug
Each for $709.50
Add to cart
 
89-949-906
View Documents
Abnova™
H00022920P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. [provided by RefSeq]

Sequence: MQGEDARYLKRKVKGGNIDVHPSEKALIVHYEVEATILGEMGDPMLGERKECQKIIRLKSLNANTDITSLARKVVEECKLIHPSKLNEVEQLLYYLQNRRDSLSGKEKKEKSSKPKDPPPFEGMEIDEVANINDMDEYIELLYEDIPDKVRGSALILQLARNPDNLEELLLNETALGALARVLREDWKQSVELATNIIYIFFCFSSFSQFHGLITHYKIGALCMNIIDHELKRHELWQEELSKKKKAVDEDPENQTLRKDYEKTFKKYQGLVVKQEQLLRVALYLLLNLAEDTRTELKMRNKNIVHMLVKALDRDNFELLILVVSFLKKLSIFMENKNDMVEMDIVEKLVKMIPCEHEDLLNITLRLLLNLSFDTGLRNKMVQVGLLPKLTALLGNDNYKQIAMCVLYHISMDDRFKSMFAYTDCIPQLMKMLFECSDERIDLELISFCINLAANKRNVQLICEGNGLKMLMKRALKFKDPLLMKMIRNISQHDGPTKNLFIDYVGDLAAQISNDEEEEFVIECLGTLANLTIPDLDWELVLKEYKLVPYLKDKLKPGAAEDDLVLEVVIMIGTVSMDDSCAALLAKSGIIPALIELLNAQQEDDEFVCQIIYVFYQMVFHQATRDVIIKETQAPAYLIDLMHDKNNEIRKVCDNTLDIIAEYDEEWAKKIQSEKFRWHNSQWLEMVESRQMDESEQYLYGDDRIEPYIHEGDILERPDLFYNSDGLIASEGAISPDFFNDYHLQNGDVVGQHSFPGSLGMDGFGQPVGILGRPATAYGFRPDEPYYYGYGS
Specifications

Specifications

KIFAP3 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
113.52kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit