Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIR2DL4 Partial ORF (NP_002246, 32 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003805Q01L

 View more versions of this product

Catalog No. 89-949-923


Add to cart

Description

Description

Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several “framework” genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response. This gene is one of the “framework” loci that is present on all haplotypes. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: FCSAWPSAVVPQGGHVTLRCHYRRGFNIFTLYKKDGVPVPELYNRIFWNSFLISPVTPAHAGTYRCRGFHPHSPTEWSAPSNPLVIMVTGLYEKP
Specifications

Specifications

NP_002246
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FCSAWPSAVVPQGGHVTLRCHYRRGFNIFTLYKKDGVPVPELYNRIFWNSFLISPVTPAHAGTYRCRGFHPHSPTEWSAPSNPLVIMVTGLYEKP
RUO
KIR2DL4
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3805
KIR2DL4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD158D|G9P|KIR103|KIR103AS
KIR2DL4
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit