Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIR3DL1 (NP_037421, 361 a.a. - 444 a.a.) Partial Recombinant Protein with His tag

Used for SDS-PAGE

Manufacturer:  Abnova™ P3516

Catalog No. 89-949-926


Add to cart

Description

Description

Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several “framework” genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response. [provided by RefSeq]

Sequence: MRGSHHHHHHGMASMTGGQQMGRDLYDDDDKDRWGSTSGTIDKLDIEFHLWCSNKKNAAVMDQEPAGNRTANSEDSDEQDPEEVTYAQLDHCVFTQRKITRPSQRPKTPPTDTILYTELPNAKPRSKVVSCP
Specifications

Specifications

Escherichia coli expression system
1 mg/mL
Liquid
3811
KIR3DL1 (Human) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 14% SDS-PAGE
MRGSHHHHHHGMASMTGGQQMGRDLYDDDDKDRWGSTSGTIDKLDIEFHLWCSNKKNAAVMDQEPAGNRTANSEDSDEQDPEEVTYAQLDHCVFTQRKITRPSQRPKTPPTDTILYTELPNAKPRSKVVSCP
RUO
KIR3DL1
Escherichia coli
His
NP_037421
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
15kDa
Escherichia coli expression system
>95% by SDS-PAGE
100 ug
Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
CD158E1|KIR|MGC119726|MGC119728|MGC126589|MGC126591|NKAT3|NKB1|NKB1B
KIR3DL1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit