Learn More
Abnova™ Human KIT (NP_000213.1, 544 a.a. - 976 a.a.) V560G mutant Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P5572
Description
This gene encodes the human homolog of the proto-oncogene c-kit. C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. This protein is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). Mutations in this gene are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous lukemia, and piebaldism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
NP_000213.1 | |
Liquid | |
76kDa | |
Baculovirus infected insect cell (Sf21) expression system | |
Loading 1 ug protein in SDS-PAGE | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C-Kit/CD117/PBT/SCFR | |
KIT | |
Baculovirus | |
GST | |
Liquid |
Functional Study, SDS-PAGE | |
3815 | |
KIT (V560G) (Human) Recombinant Protein | |
Glutathione sepharose chromatography | |
5 ug | |
RUO | |
KIT | |
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: Srctide. ATP: 1000 uM | |
Recombinant | |
Baculovirus infected insect cell (Sf21) expression system | |
87% by SDS-PAGE/CBB staining |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.