Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human KLC4 Full-length ORF (NP_958929.1, 1 a.a. - 619 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00089953P01S
Description
Sequence: MSGLVLGQRDEPAGHRLSQEEILGSTRLVSQGLEALRSEHQAVLQSLSQTIECLQQGGHEEGLVHEKARQLRRSMENIELGLSEAQVMLALASHLSTVESEKQKLRAQVRRLCQENQWLRDELAGTQQRLQRSEQAVAQLEEEKKHLEFLGQLRQYDEDGHTSEEKEGDATKDSLDDLFPNEEEEDPSNGLSRGQGATAAQQGGYEIPARLRTLHNLVIQYAAQGRYEVAVPLCKQALEDLERTSGRGHPDVATMLNILALVYRDQNKYKEAAHLLNDALSIRESTLGPDHPAVAATLNNLAVLYGKRGKYKEAEPLCQRALEIREKVLGTNHPDVAKQLNNLALLCQNQGKYEAVERYYQRALAIYEGQLGPDNPNVARTKNNLASCYLKQGKYAEAETLYKEILTRAHVQEFGSVDDDHKPIWMHAEEREEMSKSRHHEGGTPYAEYGGWYKACKVSSPTVNTTLRNLGALYRRQGKLEAAETLEECALRSRRQGTDPISQTKVAELLGESDGRRTSQEGPGDSVKFEGGEDASVAVEWSGDGSGTLQRSGSLGKIRDVLRRSSELLVRKLQGTEPRPSSSNMKRAASLNYLNQPSAAPLQVSRGLSASTMDLSSSSSpecifications
NP_958929.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KNSL8/MGC111777/bA387M24.3 | |
KLC4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
89953 | |
KLC4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSGLVLGQRDEPAGHRLSQEEILGSTRLVSQGLEALRSEHQAVLQSLSQTIECLQQGGHEEGLVHEKARQLRRSMENIELGLSEAQVMLALASHLSTVESEKQKLRAQVRRLCQENQWLRDELAGTQQRLQRSEQAVAQLEEEKKHLEFLGQLRQYDEDGHTSEEKEGDATKDSLDDLFPNEEEEDPSNGLSRGQGATAAQQGGYEIPARLRTLHNLVIQYAAQGRYEVAVPLCKQALEDLERTSGRGHPDVATMLNILALVYRDQNKYKEAAHLLNDALSIRESTLGPDHPAVAATLNNLAVLYGKRGKYKEAEPLCQRALEIREKVLGTNHPDVAKQLNNLALLCQNQGKYEAVERYYQRALAIYEGQLGPDNPNVARTKNNLASCYLKQGKYAEAETLYKEILTRAHVQEFGSVDDDHKPIWMHAEEREEMSKSRHHEGGTPYAEYGGWYKACKVSSPTVNTTLRNLGALYRRQGKLEAAETLEECALRSRRQGTDPISQTKVAELLGESDGRRTSQEGPGDSVKFEGGEDASVAVEWSGDGSGTLQRSGSLGKIRDVLRRSSELLVRKLQGTEPRPSSSNMKRAASLNYLNQPSAAPLQVSRGLSASTMDLSSSS | |
RUO | |
KLC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction