Learn More
Abnova™ Human KLF8 Full-length ORF (NP_009181.2, 1 a.a. - 359 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011279P01L
Description
This gene encodes a protein which is a member of the Sp/KLF family of transcription factors. Members of this family contain a C-terminal DNA-binding domain with three Kruppel-like zinc fingers. The encoded protein is thought to play an important role in the regulation of epithelial to mesenchymal transition, a process which occurs normally during development but also during metastasis. A pseudogene has been identified on chromosome 16. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MVDMDKLINNLEVQLNSEGGSMQVFKQVTASVRNRDPPEIEYRSNMTSPTLLDANPMENPALFNDIKIEPPEELLASDFSLPQVEPVDLSFHKPKAPLQPASMLQAPIRPPKPQSSPQTLVVSTSTSDMSTSANIPTVLTPGSVLTSSQSTGSQQILHVIHTIPSVSLPNKMGGLKTIPVVVQSLPMVYTTLPADGGPAAITVPLIGGDGKNAGSVKVDPTSMSPLEIPSDSEESTIESGSSALQSLQGLQQEPAAMAQMQGEESLDLKRRRIHQCDFAGCSKVYTKSSHLKAHRRIHTGEKPYKCTWDGCSWKFARSDELTRHFRKHTGIKPFRCTDCNRSFSRSDHLSLHRRRHDTMSpecifications
NP_009181.2 | |
Liquid | |
11279 | |
KLF8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVDMDKLINNLEVQLNSEGGSMQVFKQVTASVRNRDPPEIEYRSNMTSPTLLDANPMENPALFNDIKIEPPEELLASDFSLPQVEPVDLSFHKPKAPLQPASMLQAPIRPPKPQSSPQTLVVSTSTSDMSTSANIPTVLTPGSVLTSSQSTGSQQILHVIHTIPSVSLPNKMGGLKTIPVVVQSLPMVYTTLPADGGPAAITVPLIGGDGKNAGSVKVDPTSMSPLEIPSDSEESTIESGSSALQSLQGLQQEPAAMAQMQGEESLDLKRRRIHQCDFAGCSKVYTKSSHLKAHRRIHTGEKPYKCTWDGCSWKFARSDELTRHFRKHTGIKPFRCTDCNRSFSRSDHLSLHRRRHDTM | |
RUO | |
KLF8 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BKLF3/DKFZp686O08126/DXS741/MGC138314/ZNF741 | |
KLF8 | |
Yes | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.