Learn More
Abnova™ Human KLF8 Full-length ORF (NP_009181.2, 1 a.a. - 359 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$487.50 - $752.00
Specifications
Accession Number | NP_009181.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11279 |
Molecular Weight (g/mol) | 65.7kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-950-009
|
Abnova™
H00011279P01S |
10 ug |
Each for $487.50
|
|
|||||
89-950-010
|
Abnova™
H00011279P01L |
25 ug |
Each for $752.00
|
|
|||||
Description
This gene encodes a protein which is a member of the Sp/KLF family of transcription factors. Members of this family contain a C-terminal DNA-binding domain with three Kruppel-like zinc fingers. The encoded protein is thought to play an important role in the regulation of epithelial to mesenchymal transition, a process which occurs normally during development but also during metastasis. A pseudogene has been identified on chromosome 16. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MVDMDKLINNLEVQLNSEGGSMQVFKQVTASVRNRDPPEIEYRSNMTSPTLLDANPMENPALFNDIKIEPPEELLASDFSLPQVEPVDLSFHKPKAPLQPASMLQAPIRPPKPQSSPQTLVVSTSTSDMSTSANIPTVLTPGSVLTSSQSTGSQQILHVIHTIPSVSLPNKMGGLKTIPVVVQSLPMVYTTLPADGGPAAITVPLIGGDGKNAGSVKVDPTSMSPLEIPSDSEESTIESGSSALQSLQGLQQEPAAMAQMQGEESLDLKRRRIHQCDFAGCSKVYTKSSHLKAHRRIHTGEKPYKCTWDGCSWKFARSDELTRHFRKHTGIKPFRCTDCNRSFSRSDHLSLHRRRHDTMSpecifications
NP_009181.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVDMDKLINNLEVQLNSEGGSMQVFKQVTASVRNRDPPEIEYRSNMTSPTLLDANPMENPALFNDIKIEPPEELLASDFSLPQVEPVDLSFHKPKAPLQPASMLQAPIRPPKPQSSPQTLVVSTSTSDMSTSANIPTVLTPGSVLTSSQSTGSQQILHVIHTIPSVSLPNKMGGLKTIPVVVQSLPMVYTTLPADGGPAAITVPLIGGDGKNAGSVKVDPTSMSPLEIPSDSEESTIESGSSALQSLQGLQQEPAAMAQMQGEESLDLKRRRIHQCDFAGCSKVYTKSSHLKAHRRIHTGEKPYKCTWDGCSWKFARSDELTRHFRKHTGIKPFRCTDCNRSFSRSDHLSLHRRRHDTM | |
RUO | |
KLF8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
11279 | |
KLF8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BKLF3/DKFZp686O08126/DXS741/MGC138314/ZNF741 | |
KLF8 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.