Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KLHDC3 Partial ORF (NP_476502, 1 a.a. - 102 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00116138Q01L

 View more versions of this product

Catalog No. 89-950-016


Add to cart

Description

Description

The protein encoded by this gene contains six repeated kelch motifs that are structurally similar to recombination activating gene 2 (RAG2), a protein involved in the activation of the V(D)J recombination. In mouse, this gene is found to express specifically in testis. Its expression in pachytene spermatocytes is localized to cytoplasma and meiotic chromatin, which suggests that this gene may be involved in meiotic recombination. [provided by RefSeq]

Sequence: MLRWTVHLEGGPRRVNHAAVAVGHRVYSFGGYCSGEDYETLRQIDVHIFNAVSLRWTKLPPVKSAIRGQAPVVPYMRYGHSTVLIDDTVLLWGGRNDTEGAC
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.96kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLRWTVHLEGGPRRVNHAAVAVGHRVYSFGGYCSGEDYETLRQIDVHIFNAVSLRWTKLPPVKSAIRGQAPVVPYMRYGHSTVLIDDTVLLWGGRNDTEGAC
RUO
KLHDC3
Wheat Germ (in vitro)
GST
NP_476502
Liquid
116138
KLHDC3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PEAS|RP1-20C7.3|dJ20C7.3|hPEAS
KLHDC3
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit