Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KLK13 Full-length ORF (NP_056411.1, 1 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026085P01S

 View more versions of this product

Catalog No. 89-950-081


Add to cart

Description

Description

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Expression of this gene is regulated by steroid hormones and may be useful as a marker for breast cancer. An additional transcript variant has been identified, but its full length sequence has not been determined. [provided by RefSeq]

Sequence: MWPLALVIASLTLALSGGVSQESSKVLNTNGTSGFLPGGYTCFPHSQPWQAALLVQGRLLCGGVLVHPKWVLTAAHCLKEGLKVYLGKHALGRVEAGEQVREVVHSIPHPEYRRSPTHLNHDHDIMLLELQSPVQLTGYIQTLPLSHNNRLTPGTTCRVSGWGTTTSPQVNYPKTLQCANIQLRSDEECRQVYPGKITDNMLCAGTKEGGKDSCEGDSGGPLVCNRTLYGIVSWGDFPCGQPDRPGVYTRVSRYVLWIRETIRKYETQQQKWLKGPQ
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp586J1923|KLK-L4|KLKL4
KLK13
Yes
NP_056411.1
Liquid
26085
KLK13 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWPLALVIASLTLALSGGVSQESSKVLNTNGTSGFLPGGYTCFPHSQPWQAALLVQGRLLCGGVLVHPKWVLTAAHCLKEGLKVYLGKHALGRVEAGEQVREVVHSIPHPEYRRSPTHLNHDHDIMLLELQSPVQLTGYIQTLPLSHNNRLTPGTTCRVSGWGTTTSPQVNYPKTLQCANIQLRSDEECRQVYPGKITDNMLCAGTKEGGKDSCEGDSGGPLVCNRTLYGIVSWGDFPCGQPDRPGVYTRVSRYVLWIRETIRKYETQQQKWLKGPQ
RUO
KLK13
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit