Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KLK15 Full-length ORF (NP_075382.1, 1 a.a. - 121 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055554P01L

 View more versions of this product

Catalog No. 89-950-087


Add to cart

Description

Description

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. In prostate cancer, this gene has increased expression, which indicates its possible use as a diagnostic or prognostic marker for prostate cancer. The gene contains multiple polyadenylation sites and alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MWLLLTLSFLLASTAAQDGDKLLEGDECAPHSQPWQVALYERGRFNCGASLISPHWVLSAAHCQSRFMRVRLGEHNLRKRDGPEQLRTTSRVIPHPRYEARSHRNDIMLLRLVQPARLNPQ
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
40.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACO|HSRNASPH
KLK15
Yes
NP_075382.1
Liquid
55554
KLK15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWLLLTLSFLLASTAAQDGDKLLEGDECAPHSQPWQVALYERGRFNCGASLISPHWVLSAAHCQSRFMRVRLGEHNLRKRDGPEQLRTTSRVIPHPRYEARSHRNDIMLLRLVQPARLNPQ
RUO
KLK15
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit