Learn More
Raybiotech Inc Human Kallikrein 6 ELISA Kit
SDP
Supplier:  Raybiotech Inc ELHKLK61
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human KLK6. Other species not determined.
Catalog No. 50-194-9481