Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human KLRF1 Full-length ORF (AAH98166.1, 1 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051348P01S
Description
Sequence: MQDEERYMTLNVQSKKRSSAQTSQLTFKDYSVTLHWYKILLGISGTVNGILTLTLISLILLVSQGVLLKCQKGSCSNATQYEDTGDLKVNNGTRRNISNKDLCASRSADQTVPRPHFRLLYRKTSpecifications
AAH98166.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10μg | |
RUO | |
KLRF1 | |
Wheat Germ (in vitro) | |
GST Tag | |
Liquid |
Protein Array, ELISA, Western Blot | |
51348 | |
Human KLRF1 Full-length ORF (AAH98166.1, 1 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
CLEC5C, MGC119907, MGC119908, MGC119909 | |
KLRF1 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction