Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KNTC1 Partial ORF (NP_055523, 2100 a.a. - 2209 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009735Q01L

 View more versions of this product

Catalog No. 89-950-157


Add to cart

Description

Description

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division. Experimental evidence indicated that the encoded protein functioned in a similar manner to that of the Drosophila rough deal protein. [provided by RefSeq]

Sequence: DPQVILKQLEEHMNTGQLAGFSHQIRSLILNNIINKKEFGILAKTKYFQMLKMHAMNTNNITELVNYLANDLSLDEASVLITEYSKHCGKPVPPDTAPCEILKMFLSGLS
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DPQVILKQLEEHMNTGQLAGFSHQIRSLILNNIINKKEFGILAKTKYFQMLKMHAMNTNNITELVNYLANDLSLDEASVLITEYSKHCGKPVPPDTAPCEILKMFLSGLS
RUO
KNTC1
Wheat Germ (in vitro)
GST
NP_055523
Liquid
9735
KNTC1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ36151|KIAA0166|ROD
KNTC1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit