Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KPNA6 Full-length ORF (NP_036448.1, 1 a.a. - 536 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name KPNA6 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 86.4kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-950-174
View Documents
Abnova™
H00023633P01S
10 ug
Each for $460.50
Add to cart
 
89-950-175
View Documents
Abnova™
H00023633P01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal (NLS) requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. The protein encoded by this gene is a member of the importin alpha family. [provided by RefSeq]

Sequence: METMASPGKDNYRMKSYKNNALNPEEMRRRREEEGIQLRKQKREQQLFKRRNVELINEEAAMFDSLLMDSYVSSTTGESVITREMVEMLFSDDSDLQLATTQKFRKLLSKEPSPPIDEVINTPRVVDRFVEFLKRNENCTLQFEAAWALTNIASGTSQQTKIVIEAGAVPIFIELLNSDFEDVQEQAVWALGNIAGDSSVCRDYVLNCSILNPLLTLLTKSTRLTMTRNAVWALSNLCRGKNPPPEFAKVSPCLPVLSRLLFSSDSDLLADACWALSYLSDGPNEKIQAVIDSGVCRRLVELLMHNDYKVASPALRAVGNIVTGDDIQTQVILNCSALPCLLHLLSSPKESIRKEACWTISNITAGNRAQIQAVIDANIFPVLIEILQKAEFRTRKEAAWAITNATSGGTPEQIRYLVSLGCIKPLCDLLTVMDSKIVQVALNGLENILRLGEQEGKRSGSGVNPYCGLIEEAYGLDKIEFLQSHENQEIYQKAFDLIEHYFGVEDDDSSLAPQVDETQQQFIFQQPEAPMEGFQL
Specifications

Specifications

KPNA6 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
86.4kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit