Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kremen-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Kremen-2 |
---|---|
Clone | 276709 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Kremen-2 Monoclonal specifically detects Kremen-2 in Human samples. It is validated for Western Blot.Specifications
Kremen-2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8NCW0 | |
79412 | |
Mouse myeloma cell line NS0-derived recombinant human Kremen-2 Gln19-Ala364 Accession # Q8NCW0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Kremen-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Kremen-1 or recombinant mouse Kremen-2 is observed. |
276709 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Dickkopf receptor 2, kremen protein 2, Kremen2, kringle containing transmembrane protein 2, Kringle domain-containing transmembrane protein 2, KRM2Kringle-containing protein marking the eye and the nose, MGC10791, MGC16709 | |
KREMEN2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title