Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRT15 Full-length ORF (NP_002266.2, 1 a.a. - 456 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003866P02L

 View more versions of this product

Catalog No. 89-950-208


Add to cart

Description

Description

The protein encoded by this gene is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21.2. [provided by RefSeq]

Sequence: MTTTFLQTSSSTFGGGSTRGGSLLAGGGGFGGGSLSGGGGSRSISASSARFVSSGSGGGYGGGMRVCGFGGGAGSVFGGGFGGGVGGGFGGGFGGGDGGLLSGNEKITMQNLNDRLASYLDKVRALEEANADLEVKIHDWYQKQTPTSPECDYSQYFKTIEELRDKIMATTIDNSRVILEIDNARLAADDFRLKYENELALRQGVEADINGLRRVLDELTLARTDLEMQIEGLNEELAYLKKNHEEEMKEFSSQLAGQVNVEMDAAPGVDLTRVLAEMREQYEAMAEKNRRDVEAWFFSKTEELNKEVASNTEMIQTSKTEITDLRRTMQELEIELQSQLSMKAGLENSLAETECRYATQLQQIQGLIGGLEAQLSELRCEMEAQNQEYKMLLDIKTRLEQEIATYRSLLEGQDAKMAGIGIREASSGGGGSSSNFHINVEESVDGQVVSSHKREI
Specifications

Specifications

NP_002266.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CK15|K15|K1CO
KRT15
Yes
wheat germ expression system
Liquid
3866
KRT15 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue
MTTTFLQTSSSTFGGGSTRGGSLLAGGGGFGGGSLSGGGGSRSISASSARFVSSGSGGGYGGGMRVCGFGGGAGSVFGGGFGGGVGGGFGGGFGGGDGGLLSGNEKITMQNLNDRLASYLDKVRALEEANADLEVKIHDWYQKQTPTSPECDYSQYFKTIEELRDKIMATTIDNSRVILEIDNARLAADDFRLKYENELALRQGVEADINGLRRVLDELTLARTDLEMQIEGLNEELAYLKKNHEEEMKEFSSQLAGQVNVEMDAAPGVDLTRVLAEMREQYEAMAEKNRRDVEAWFFSKTEELNKEVASNTEMIQTSKTEITDLRRTMQELEIELQSQLSMKAGLENSLAETECRYATQLQQIQGLIGGLEAQLSELRCEMEAQNQEYKMLLDIKTRLEQEIATYRSLLEGQDAKMAGIGIREASSGGGGSSSNFHINVEESVDGQVVSSHKREI
RUO
KRT15
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit