Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRT35 Full-length ORF (AAI46509.1, 1 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003886P01L

 View more versions of this product

Catalog No. 89-950-291


Add to cart

Description

Description

The protein encoded by this gene is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq]

Sequence: MASKCLKAGFSSGSLKSPGGASGGSTRVSAMYSSSSCKLPSLSPVARSFSACSVGLGRSSYRATSCLPALCLPAGGFATSYSGGGGWFGEGILTGNEKETMQSLNDRLAGYLEKVRQLEQENASLESRIREWCEQQVPYMCPDYQSYFRTIEELQKKTLCSKAENARLVVEIDNAKLAADDFRTKYETEVSLRQLVESDINGLRRILDDLTLCKSDLEAQVESLKEELLCLKKNHEEEVNSLRCQLGDRLNVEVDAAPPVDLNRVLEEMRCQYETLVENNRRDAEDWLDTQSEELNQQVVSSSEQLQSCQAEIIELRRTVNALEIELQAQHSMRDALESTLAETEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLECEINTYRGLLESEDSKLPCNPCAPDYSPSKSCLPCLPAASCGPSAARTNCSPRPICVPCPGGRF
Specifications

Specifications

AAI46509.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HA5|Ha-5|KRTHA5|hHa5
KRT35
Yes
wheat germ expression system
Liquid
3886
KRT35 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASKCLKAGFSSGSLKSPGGASGGSTRVSAMYSSSSCKLPSLSPVARSFSACSVGLGRSSYRATSCLPALCLPAGGFATSYSGGGGWFGEGILTGNEKETMQSLNDRLAGYLEKVRQLEQENASLESRIREWCEQQVPYMCPDYQSYFRTIEELQKKTLCSKAENARLVVEIDNAKLAADDFRTKYETEVSLRQLVESDINGLRRILDDLTLCKSDLEAQVESLKEELLCLKKNHEEEVNSLRCQLGDRLNVEVDAAPPVDLNRVLEEMRCQYETLVENNRRDAEDWLDTQSEELNQQVVSSSEQLQSCQAEIIELRRTVNALEIELQAQHSMRDALESTLAETEARYSSQLAQMQCMITNVEAQLAEIRADLERQNQEYQVLLDVRARLECEINTYRGLLESEDSKLPCNPCAPDYSPSKSCLPCLPAASCGPSAARTNCSPRPICVPCPGGRF
RUO
KRT35
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit