Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRT75 Full-length ORF (AAI60175.1, 1 a.a. - 551 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009119P01L

 View more versions of this product

Catalog No. 89-950-249


Add to cart

Description

Description

This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. This gene is expressed in the companion layer, upper germinative matrix region of the hair follicle, and medulla of the hair shaft. The encoded protein plays an essential role in hair and nail formation. Variations in this gene have been associated with the hair disorders pseudofolliculitis barbae (PFB) and loose anagen hair syndrome (LAHS). [provided by RefSeq]

Sequence: MSRQSSITFQSGSRRGFSTTSAITPAAGRSRFSSVSVARSAAGSGGLGRISSAGASFGSRSLYNLGGAKRVSINGCGSSCRSGFGGRASNRFGVNSGFGYGGGVGGGFSGPSFPVCPPGGIQEVTVNQSLLTPLHLQIDPTIQRVRAEEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGSRTVRQNLEPLFDSYTSELRRQLESITTERGRLEAELRNMQDVVEDFKVRYEDEINKRTAAENEFVALKKDVDAAYMNKVELEAKVKSLPEEINFIHSVFDAELSQLQTQVGDTSVVLSMDNNRNLDLDSIIAEVKAQYEDIANRSRAEAESWYQTKYEELQVTAGRHGDDLRNTKQEISEMNRMIQRLRAEIDSVKKQCSSLQTAIADAEQRGELALKDARAKLVDLEEALQKAKQDMARLLREYQELMNIKLALDVEIATYRKLLEGEECRLSGEGVSPVNISVVTSTLSSGYGSGSSIGGGNLGLGGGSGYSFTTSGGHSLGAGLGGSGFSATSNRGLGGSGSSVKFVSTTSSSQKSYTH
Specifications

Specifications

AAI60175.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
K6HF|KB18|PFB
KRT75
Yes
wheat germ expression system
Liquid
9119
KRT75 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MSRQSSITFQSGSRRGFSTTSAITPAAGRSRFSSVSVARSAAGSGGLGRISSAGASFGSRSLYNLGGAKRVSINGCGSSCRSGFGGRASNRFGVNSGFGYGGGVGGGFSGPSFPVCPPGGIQEVTVNQSLLTPLHLQIDPTIQRVRAEEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGSRTVRQNLEPLFDSYTSELRRQLESITTERGRLEAELRNMQDVVEDFKVRYEDEINKRTAAENEFVALKKDVDAAYMNKVELEAKVKSLPEEINFIHSVFDAELSQLQTQVGDTSVVLSMDNNRNLDLDSIIAEVKAQYEDIANRSRAEAESWYQTKYEELQVTAGRHGDDLRNTKQEISEMNRMIQRLRAEIDSVKKQCSSLQTAIADAEQRGELALKDARAKLVDLEEALQKAKQDMARLLREYQELMNIKLALDVEIATYRKLLEGEECRLSGEGVSPVNISVVTSTLSSGYGSGSSIGGGNLGLGGGSGYSFTTSGGHSLGAGLGGSGFSATSNRGLGGSGSSVKFVSTTSSSQKSYTH
RUO
KRT75
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit