Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRTAP13-3 Full-length ORF (NP_853653.1, 1 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00337960P01S

 View more versions of this product

Catalog No. 89-950-268


Add to cart

Description

Description

Sequence: MSYNCCSRNFSSCSHGGYLHYPGSSCGSSYPSNLVYSTDLCSPSTCQLGSSLYRGCQETCWRPNSCQTLCVESSPCHTSCYYPRTHMLCNSCLTMHVGSRGFGSNSCCSLSCGSRSCSSLGCGSNGFRYLNYRIHTSPSQSYRSRFCHPIYFPPRRWFHSSCYQPFCRSGFY
Specifications

Specifications

NP_853653.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KAP13.3|MGC119828
KRTAP13-3
Yes
wheat germ expression system
Liquid
337960
KRTAP13-3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSYNCCSRNFSSCSHGGYLHYPGSSCGSSYPSNLVYSTDLCSPSTCQLGSSLYRGCQETCWRPNSCQTLCVESSPCHTSCYYPRTHMLCNSCLTMHVGSRGFGSNSCCSLSCGSRSCSSLGCGSNGFRYLNYRIHTSPSQSYRSRFCHPIYFPPRRWFHSSCYQPFCRSGFY
RUO
KRTAP13-3
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit