Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRTAP21-2 (NM_181617) Full-length Recombinant Protein expressed in yeast

Used for SDS-PAGE

Manufacturer:  Abnova™ P3777

Catalog No. 89-950-274


Add to cart

Description

Description

Sequence: MCCNYYRNCCGGCGYGSGWSSGCGYGCGYGCGYGSGCRYGSGYGTGCGYGCGYGSGCGYGCGYSSSCCGYRPLCYRRCYSSCY
Specifications

Specifications

NM_181617
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
8.555kDa
Yeast expression system
0.9
MCCNYYRNCCGGCGYGSGWSSGCGYGCGYGCGYGSGCRYGSGYGTGCGYGCGYGSGCGYGCGYSSSCCGYRPLCYRRCYSSCY
RUO
KRTAP21-2
Yeast
None
Yeast expression system
Liquid
337978
KRTAP21-2 (Human) Recombinant Protein
Affinity Purification
100 ug
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
KAP21.2
KRTAP21-2
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit