Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRTAP3-3 Full-length ORF (NP_149441.1, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00085293P01S

 View more versions of this product

Catalog No. 89-950-275


Add to cart

Description

Description

This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the high sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21. [provided by RefSeq]

Sequence: MDCCASRGCSVPTGPATTICSSDKSCRCGVCLPSTCPHTVWLLEPTCCDNCPPPCHIPQPCVPTCFLLNSCQPTPGLETLNLTTFTQPCCEPCLPRGC
Specifications

Specifications

NP_149441.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KAP3.3|KRTAP3.3|MGC95374
KRTAP3-3
Yes
wheat germ expression system
Liquid
85293
KRTAP3-3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDCCASRGCSVPTGPATTICSSDKSCRCGVCLPSTCPHTVWLLEPTCCDNCPPPCHIPQPCVPTCFLLNSCQPTPGLETLNLTTFTQPCCEPCLPRGC
RUO
KRTAP3-3
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit