Learn More
Raybiotech IncSupplier Diversity Partner Human L-Selectin (CD62L) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHLSELECTIN1
For cell culture supernatants, plasma, and serum samples. Sensitivity: 100 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, I-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MIP-1, MIP-1 beta, MIP-1delta, PARC, RANTES, SCF, TARC, TGF-beta 1, TGF-beta 2, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Catalog No. 50-194-9511