Learn More
Raybiotech IncSupplier Diversity Partner Human L1CAM (NCAM-L1) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHL1CAM1
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.8 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes human L1CAM. Other species not yet determined.
Catalog No. 50-194-9485