Learn More
Abnova™ Human L3MBTL Full-length ORF (AAH39820, 1 a.a. - 772 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00026013P01S
Description
This gene encodes the homolog of a protein identified in Drosophila as a suppressor of malignant transformation of neuroblasts and ganglion-mother cells in the optic centers of the brain. This gene product is localized to condensed chromosomes in mitotic cells. Overexpression of this gene in a glioma cell line results in improper nuclear segregation and cytokinesis producing multinucleated cells. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MRRREGHGTDSEMGQGPVRESQSSDPPALQFRISEYKPLNMAGVEQPPTPELRQEGVTEYEDGGAPAGDGEAGPQQAEDHPQNPPEDPNQDPPEDDSTCQCQACGPHQAAGPDLGSSNDGCPQLFQERSVIVENSSGSTSASELLKPMKKRKRREYQSPSEEESEPEAMEKQEEGKDPEGQPTASTPESEEWSSSQPATGEKKECWSWESYLEEQKAITAPVSLFQDSQAVTHNKNGFKLGMKLEGIDPQHPSMYFILTVAEVCGYRLRLHFDGYSECHDFWVNANSPDIHPAGWFEKTGHKLQPPKGYKEEEFSWSQYLRSTRAQAAPKHLFVSQSHSPPPLGFQVGMKLEAVDRMNPSLVCVASVTDVVDSRFLVHFDNWDDTYDYWCDPSSPYIHPVGWCQKQGKPLTPPQDYPDPDNFCWEKYLEETGASAVPTWAFKVRPPHSFLVNMKLEAVDRRNPALIRVASVEDVEDHRIKIHFDGWSHGYDFWIDADHPDIHPAGWCSKTGHPLQPPLGPREPSSASPGGCPPLSYRSLPHTRTSKYSFHHRKCPTPGCDGSGHVTGKFTAHHCLSGCPLAERNQSRLKAELSDSEASARKKNLSGFSPRKKPRHHGRIGRPPKYRKIPQEDFQTLTPDVVHQSLFMSALSAHPDRSLSVCWEQHCKLLPGVAGISASTVAKWTIDEVFGFVQTLTGCEDQARLFKDEARIVRVTHVSGKTLVWTVAQLGDLVCSDHLQEGKGILETGVHSLLCSLPTHLLAKLSFASDSQYSpecifications
AAH39820 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.66kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586P1522/FLJ41181/H-L(3)MBT/KIAA0681/L3MBTL1/dJ138B7.3 | |
L3MBTL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26013 | |
L3MBTL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRREGHGTDSEMGQGPVRESQSSDPPALQFRISEYKPLNMAGVEQPPTPELRQEGVTEYEDGGAPAGDGEAGPQQAEDHPQNPPEDPNQDPPEDDSTCQCQACGPHQAAGPDLGSSNDGCPQLFQERSVIVENSSGSTSASELLKPMKKRKRREYQSPSEEESEPEAMEKQEEGKDPEGQPTASTPESEEWSSSQPATGEKKECWSWESYLEEQKAITAPVSLFQDSQAVTHNKNGFKLGMKLEGIDPQHPSMYFILTVAEVCGYRLRLHFDGYSECHDFWVNANSPDIHPAGWFEKTGHKLQPPKGYKEEEFSWSQYLRSTRAQAAPKHLFVSQSHSPPPLGFQVGMKLEAVDRMNPSLVCVASVTDVVDSRFLVHFDNWDDTYDYWCDPSSPYIHPVGWCQKQGKPLTPPQDYPDPDNFCWEKYLEETGASAVPTWAFKVRPPHSFLVNMKLEAVDRRNPALIRVASVEDVEDHRIKIHFDGWSHGYDFWIDADHPDIHPAGWCSKTGHPLQPPLGPREPSSASPGGCPPLSYRSLPHTRTSKYSFHHRKCPTPGCDGSGHVTGKFTAHHCLSGCPLAERNQSRLKAELSDSEASARKKNLSGFSPRKKPRHHGRIGRPPKYRKIPQEDFQTLTPDVVHQSLFMSALSAHPDRSLSVCWEQHCKLLPGVAGISASTVAKWTIDEVFGFVQTLTGCEDQARLFKDEARIVRVTHVSGKTLVWTVAQLGDLVCSDHLQEGKGILETGVHSLLCSLPTHLLAKLSFASDSQY | |
RUO | |
L3MBTL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.