Learn More
Raybiotech IncSupplier Diversity Partner L-Series Human Antibody Array 2000 Membrane Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner AAHBLM2000B2
Detects 2000 Human Proteins. Suitable for cell culture supernatants (A) and serum, plasma and tissue lysates (B). Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAH-BLM-2000B-2
Catalog No. 50-194-8509