Learn More
Raybiotech IncSupplier Diversity Partner L-Series Human Antibody Array 1000 Glass Slide Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner AAHBLG10008
Detects 1000 Human Proteins. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAH-BLG-1000-8
Catalog No. 50-194-8490