Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Lactate Dehydrogenase A/LDHA Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$286.52 - $503.44
Specifications
Accession Number | P00338 |
---|---|
For Use With (Application) | Enzyme Activity |
Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
Gene ID (Entrez) | 3939 |
Molecular Weight (g/mol) | M.W. (Observed): 36 kDa, reducing conditions; M.W. (theoretical): 38 kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
91-58H-A050
|
R&D Systems™
9158HA050 |
50 μg |
Each for $503.44
|
|
|||||
91-58H-A010
|
R&D Systems™
9158HA010 |
10 μg |
Each for $286.52
|
|
|||||
Description
Measured by its ability to reduce pyruvate to lactate. The specific activity is >95,000 pmol/min/μg, as measured under the described conditions.Specifications
P00338 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 36 kDa, reducing conditions; M.W. (theoretical): 38 kDa | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDHA, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59 | |
Measured by its ability to reduce pyruvate to lactate. The specific activity is >95,000 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com. | |
Human | |
E. coli-derived Ala2-Phe332, with N-terminal Met and 6-His tag | |
Lactate Dehydrogenase A/LDHA |
Enzyme Activity | |
3939 | |
E. coli-derived human Lactate Dehydrogenase A/LDHA protein Ala2-Phe332, with N-terminal Met and 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Lactate Dehydrogenase A/LDHA | |
Unconjugated | |
Recombinant | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title