Learn More
Abnova™ Human LAMA2 Partial ORF (NP_000417, 3013 a.a. - 3122 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_000417 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3908 |
Molecular Weight (g/mol) | 37.84kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-950-345
|
Abnova™
H00003908Q01S |
10 ug |
Each for $499.00
|
|
|||||
89-950-346
|
Abnova™
H00003908Q01L |
25 ug |
Each for $769.00
|
|
|||||
Description
Laminin, an extracellular protein, is a major component of the basement membrane. It is thought to mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. It is composed of three subunits, alpha, beta, and gamma, which are bound to each other by disulfide bonds into a cross-shaped molecule. This gene encodes the alpha 2 chain, which constitutes one of the subunits of laminin 2 (merosin) and laminin 4 (s-merosin). Mutations in this gene have been identified as the cause of congenital merosin-deficient muscular dystrophy. Two transcript variants encoding different proteins have been found for this gene. [provided by RefSeq]
Sequence: DAGVPGHLCDGQWHKVTANKIKHRIELTVDGNQVEAQSPNPASTSADTNDPVFVGGFPDDLKQFGLTTSIPFRGCIRSLKLTKGTGKPLEVNFAKALELRGVQPVSCPANSpecifications
NP_000417 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LAMM | |
LAMA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3908 | |
LAMA2 (Human) Recombinant Protein (Q01) | |
DAGVPGHLCDGQWHKVTANKIKHRIELTVDGNQVEAQSPNPASTSADTNDPVFVGGFPDDLKQFGLTTSIPFRGCIRSLKLTKGTGKPLEVNFAKALELRGVQPVSCPAN | |
RUO | |
LAMA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.