Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LAMA5 Full-length ORF (AAH03355.1, 1 a.a. - 670 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003911P01

Catalog No. 89-950-354


Add to cart

Description

Description

Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined. [provided by RefSeq]

Sequence: MGVSLRDKKVHWVYQLGEAGPAVLSIDEDIGEQFAAVSLDRTLQFGHMSVTVERQMIQETKGDTVAPGAEGLLNLRPDDFVFYVGGYPSTFTPPPLLRFPGYRGCIEMDTLNEEVVSLYNFERTFQLDTAVDRPCARSKSTGDPWLTDGSYLDGTGFARISFDSQISTTKRFEQELRLVSYSGVLFFLKQQIQVFLLGGSRKRVLVRVERATVYSVEQDNDLELADAYYLGGVPPDQLPPSLRRLFPTGGSVRGCVKGIKALGKYVDLKRLNTTGVSAGCTADLLVGRAMTFHGHGFLRLALSNVAPLTGNVYSGFGFHSAQDSALLYYRASPDGLCQVSLQQGRVSLQLLRTEVKTQAGFADGAPHYVAFYSNATGVWLYVDDQLQQMKPHRGPPPELQPQPEGPPRLLLGGLPESGTIYNFSGCISNVFVQRLLGPQRVFDLQQNLGSVNVSTGCAPALQAQTPGLGPRGLQATARKASRRSRQPARHPACMLPPHLRTTRDSYQFGGSLSSHLEFVGILARHRNWPSLSMHVLPRSSRGLLLFTARLRPGSPSLALFLSNGHFVAQMEGLGTRLRAQSRQRSRPGRWHKVSVRWEKNRILLVTDGARAWSQEGPHRQHQGAEHPQPHTLFVGGLPASSHSSKLPTSQELHCLMWAWNWRCGPWQSPD
Specifications

Specifications

AAH03355.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
100.4kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1907
LAMA5
Yes
wheat germ expression system
Liquid
3911
LAMA5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGVSLRDKKVHWVYQLGEAGPAVLSIDEDIGEQFAAVSLDRTLQFGHMSVTVERQMIQETKGDTVAPGAEGLLNLRPDDFVFYVGGYPSTFTPPPLLRFPGYRGCIEMDTLNEEVVSLYNFERTFQLDTAVDRPCARSKSTGDPWLTDGSYLDGTGFARISFDSQISTTKRFEQELRLVSYSGVLFFLKQQIQVFLLGGSRKRVLVRVERATVYSVEQDNDLELADAYYLGGVPPDQLPPSLRRLFPTGGSVRGCVKGIKALGKYVDLKRLNTTGVSAGCTADLLVGRAMTFHGHGFLRLALSNVAPLTGNVYSGFGFHSAQDSALLYYRASPDGLCQVSLQQGRVSLQLLRTEVKTQAGFADGAPHYVAFYSNATGVWLYVDDQLQQMKPHRGPPPELQPQPEGPPRLLLGGLPESGTIYNFSGCISNVFVQRLLGPQRVFDLQQNLGSVNVSTGCAPALQAQTPGLGPRGLQATARKASRRSRQPARHPACMLPPHLRTTRDSYQFGGSLSSHLEFVGILARHRNWPSLSMHVLPRSSRGLLLFTARLRPGSPSLALFLSNGHFVAQMEGLGTRLRAQSRQRSRPGRWHKVSVRWEKNRILLVTDGARAWSQEGPHRQHQGAEHPQPHTLFVGGLPASSHSSKLPTSQELHCLMWAWNWRCGPWQSPD
RUO
LAMA5
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit